missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: 2-Leg Stoppers

Product Code. 10021492 Shop All Wheaton Science Products Products
Change view
Click to view available options
Product Type:
2-Leg Lyophilization Stopper
Straight Plug Stopper
Material:
Butyl Rubber
Chlorobutyl Isoprene Blend
Rubber
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Material unitSize
10021492 Straight Plug Stopper Rubber Pack of 1000
10452242 2-Leg Lyophilization Stopper Chlorobutyl Isoprene Blend Pack of 1000
11764189 2-Leg Lyophilization Stopper Butyl Rubber Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10021492 Supplier DWK Life Sciences Supplier No. 224100172
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provide airtight covers

  • Flange-type, straight plug stoppers are secured with aluminum crimp seals (06-406-11B)
  • Three-prong plug stoppers are for freeze-dry applications (06-447E, 06-451-56)
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 1.3 mm
Diameter (Metric) 20 mm
For Use With (Application) Not for lyophilization
For Use With (Equipment) Fits 1.3mm I.D., 20mm O.D. mouth of serum bottles and vials
Hardness 45 Durometer
Autoclavable Autoclavable
Color Red
Diameter (Metric) Bottom 13 mm
Material Rubber
Product Type Straight Plug Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.