missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Snap-on Stopper

Product Code. 11571105 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Gray
Red
Material:
Chlorobutyl Rubber
Rubber
Diameter (Metric):
13 mm
20 mm
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Diameter (Metric) unitSize
11571105 Gray Chlorobutyl Rubber 20 mm Pack of 1000
12492621 Red Rubber 13 mm Pack of 1000
12402641 Red Rubber 20 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11571105 Supplier DWK Life Sciences Supplier No. W224100181

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide airtight covers

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 13 mm
Diameter (Metric) 20 mm
For Use With (Equipment) Serum bottles and vials
Hardness 45 Durometer
Autoclavable Autoclavable
Color Gray
Material Chlorobutyl Rubber
Product Type Plug Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.