missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper
Provide airtight covers
£231.00 - £820.00
Specifications
Autoclavable | Yes |
---|---|
Color | Red |
Material | Rubber |
Product Type | Sleeve Stopper |
For Use With (Application) | Multi-puncture applications |
Product Code | Brand | Color | Material | For Use With (Equipment) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Material | For Use With (Equipment) | Price | Quantity & Availability | |||
11798919
|
DWK Life Sciences
224100-060 |
Red | Rubber | Fits 0.7cm I.D., 1.3cm O.D. mouth of serum bottles and vials |
£565.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
11718929
|
DWK Life Sciences
224100-020 |
Red | Rubber | Fits 0.5cm I.D., 1.1cm O.D. mouth of serum bottles and vials |
£231.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
12442641
|
DWK Life Sciences
224100-320 |
Red | Rubber | Fits 1.5cm I.D., 3cm O.D. mouth of serum bottles and vials |
£820.00
Pack of 1000 |
Estimated Shipment: 19-06-2024
Log in to see stock. |
|||
11708929
|
DWK Life Sciences
224100-161 |
Red | Rubber | Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials |
£515.00
Pack of 1000 |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||
Description
- Flange-type stoppers are secured with aluminum crimp seals
- Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications
Yes | |
Rubber | |
Multi-puncture applications |
Red | |
Sleeve Stopper | |
40 Durometer |