missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Sleeve Stopper

Product Code. 11708929 Shop All Wheaton Science Products Products
Change view
Click to view available options
Diameter (Metric) Outer:
11 mm
13 mm
20 mm
30 mm
Diameter (Metric) Inner:
5 mm
7 mm
13 mm
15 mm
Unit Size:
Pack of 1000
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outer Diameter (Metric) Inner unitSize
11708929 20 mm 13 mm Pack of 1000
11798919 13 mm 7 mm Pack of 1000
11718929 11 mm 5 mm Pack of 1000
12442641 30 mm 15 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11708929 Supplier DWK Life Sciences Supplier No. 224100161

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Provide airtight covers

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 13 mm
For Use With (Application) Multi-puncture applications
For Use With (Equipment) Fits 1.3cm I.D., 2cm O.D. mouth of serum bottles and vials
Hardness 40 Durometer
Autoclavable Autoclavable
Color Red
Diameter (Metric) Bottom 13 mm
Diameter (Metric) Outer 20 mm
Material Rubber
Product Type Sleeve Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.