missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Straight Plug Stopper

Product Code. 10036671 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Gray
Red
Material:
Butyl Rubber
Isoprene Rubber
Rubber
Diameter (Metric):
13 mm
30 mm
20 cm
Unit Size:
Pack of 1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material Diameter (Metric) unitSize
10036671 Red Rubber 30 mm Pack of 1000
12811144 Gray Butyl Rubber 20 cm Pack of 1000
11788919 Gray Isoprene Rubber 13 mm Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10036671 Supplier DWK Life Sciences Supplier No. 224100330

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Provide airtight covers

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 15 mm
Diameter (Metric) 30 mm
For Use With (Equipment) Serum bottles and vials
Hardness 40 Durometer
Autoclavable Autoclavable
Color Red
Diameter (Metric) Bottom 15 mm
Material Rubber
Product Type Straight Plug Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.