missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with Open Top

Product Code. 10015254 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10015254 Black Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10015254 Supplier DWK Life Sciences Supplier No. W240509
Add to basket
Add to basket
This item is not returnable. View return policy

Autoclavable screw caps, without liner

  • Autoclavable
  • Open top caps provide access with a syringe
TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic
Liner Material N/A
Product Type Screw Cap
For Use With (Equipment) Vacule Ampules
Closure Size 15-425
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.