Learn More
DWK Life Sciences Wheaton™ Stopper Leash For B.O.D. Bottles
Shop All Wheaton Science Products ProductsDescription
- 10mm (0.4 in.) holes at either end expand to fit securely around both stoppers and bottle necks
- Keeps stopper secured to the neck of the bottle
Specifications
Specifications
| For Use With (Equipment) | Stoppers and bottle necks |
| Material | Rubber |