missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Stainless-Steel Shaft Assembly Kit For Celstir

Product Code. 11531295 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
1000mL Flask
125mL Flask
250mL Flask
25mL Flask
6000mL Flask
8000mL Flask
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
11531295 125mL Flask Each
11551295 250mL Flask Each
11511295 25mL Flask Each
10553062 1000mL Flask Each
11571295 6000mL Flask Each
11581295 8000mL Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 11531295 Supplier DWK Life Sciences Supplier No. 356878
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Adjustable paddle blade impeller for better mixing

  • Provide maximum surface interface between culture and flask atmosphere
  • Ideal for microcarrier and suspension cultures such as insect cells, hybridomas, and adapted cell lines
  • Made from Wheaton 33 low-extractable borosilicate glass that conforms to ASTM™ Type I, Class A and USP Type I requirements
  • Impeller rod does not protrude up through top cap
  • Sidearms for 500mL to 8L flask allow vented caps which enables the flask to breathe while protecting from contamination
  • 1:1 headspace for good gas exchange
  • No contamination from stirring rod
  • Bottom dimple on 125mL and larger flask improves circulation and reduces accumulation of cells in center of the flask
  • Sizes 125 to 1000mL are graduated in 50mL increments; sizes 3000 to 8000mL are graduated in 500mL increments; sizes 25mL and 50mL are non-graduated
  • Completely autoclavable at 121°C for 20 minutes

Order Info

Assemblies 500mL and larger can be used with the vented cap (Cat. No. 22-288-526)

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 125mL Flask
Type Shaft Assembly Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.