missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Stainless-Steel Shaft Assembly Kit For Celstir
Shop All Wheaton Science Products ProductsDescription
- Provide maximum surface interface between culture and flask atmosphere
- Ideal for microcarrier and suspension cultures such as insect cells, hybridomas, and adapted cell lines
- Made from Wheaton 33 low-extractable borosilicate glass that conforms to ASTM™ Type I, Class A and USP Type I requirements
- Impeller rod does not protrude up through top cap
- Sidearms for 500mL to 8L flask allow vented caps which enables the flask to breathe while protecting from contamination
- 1:1 headspace for good gas exchange
- No contamination from stirring rod
- Bottom dimple on 125mL and larger flask improves circulation and reduces accumulation of cells in center of the flask
- Sizes 125 to 1000mL are graduated in 50mL increments; sizes 3000 to 8000mL are graduated in 500mL increments; sizes 25mL and 50mL are non-graduated
- Completely autoclavable at 121°C for 20 minutes
Order Info
Assemblies 500mL and larger can be used with the vented cap (Cat. No. 22-288-526)
Specifications
Specifications
| For Use With (Equipment) | 25mL Flask |
| Type | Shaft Assembly Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?