missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Serum Bottles and Vials

Product Code. 10071392 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
2 mL
5 mL
10 mL
20 mL
30 mL
50 mL
60 mL
100 mL
125 mL
Color:
Amber
Clear
Unit Size:
Pack of 144
Pack of 288
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Color unitSize
10071392 30 mL Amber Pack of 288
11591165 2 mL Clear Pack of 288
10520853 5 mL Clear Pack of 288
10424292 5 mL Amber Pack of 288
10768134 10 mL Clear Pack of 288
10434292 10 mL Amber Pack of 288
10453712 20 mL Clear Pack of 288
10600173 20 mL Amber Pack of 288
10632973 30 mL Clear Pack of 288
10464672 50 mL Amber Pack of 288
10642973 50 mL Clear Pack of 288
10235262 60 mL Clear Pack of 144
10011422 100 mL Amber Pack of 144
10021362 100 mL Clear Pack of 144
10071382 125 mL Clear Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 10071392 Supplier DWK Life Sciences Supplier No. 223763
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

These bottles have exceptional wall strength and superior heat and chemical tolerances

Ideal for long- and short-term sample storage and vaccine/injectable drug containers.

  • Clear borosilicate glass conforms to USP Type 1 requirements
  • Amber borosilicate glass conforms to USP Type 1 requirements
  • Shrink wrapped partitioned modules reduce breakage

Compliance

Meet requirements for Class A of the United States Pharmacopeia for Type I glass.

TRUSTED_SUSTAINABILITY

Specifications

Capacity 1.01 oz., 30 mL
Capacity (Metric) 30 mL
Capacity (English) 1.01 oz.
Color Amber
Diameter (English) 1.45 in.
Material Borosilicate Glass
Height (Metric) 62 mm
Height (English) 2.44 in.
Diameter (Metric) 37 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.