missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps With PTFE Faced Foamed Polyethylene Liner
Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
15-425
Unit Size:
Pack of 144
Description
- Can withstand autoclaving / sterilization
Specifications
Specifications
| Color | White |
| Material | Polypropylene |
| No. per Case | 144 Cs. |
| Liner Material | PTFE/Polyethylene |
| Neck Finish | 13-425 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |
| For Use With (Equipment) | Glass bottle or vial |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction