missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Round Translucent LDPE Bottles

Product Code. 12402581 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
7 mL
15 mL
30 mL
Neck Finish:
15-415
20-410
Unit Size:
Pack of 144
Pack of 72
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Neck Finish unitSize
12402581 7 mL 15-415 Pack of 144
10313442 15 mL 15-415 Pack of 144
12412581 30 mL 20-410 Pack of 72
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12402581 Supplier DWK Life Sciences Supplier No. 221140
Only null left
Add to basket
Edge
Add to basket

Very flexible, high impact strength plastic

Narrow mouth is ideal for liquids.

  • Excellent for mild and strong buffers, good chemical resistance
  • Pre-attached white polypropylene screw cap with foamed polyethylene liner

TRUSTED_SUSTAINABILITY

Specifications

Capacity 0.24 oz., 7 mL
Capacity (Metric) 7 mL
Capacity (English) 0.24 oz.
Color Translucent
Diameter (English) 0.74 in.
Material LDPE
Closure Color White
Neck Finish 15-415
Certifications/Compliance ISO 9001
Closure Included Yes
Height (Metric) 63 mm
Height (English) 2.4 in.
Closure Liner Polyethylene Foam
Closure Material Polyethylene
Closure Size 15-415 mm
Diameter (Metric) 19 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.