Learn More
DWK Life Sciences Wheaton™ PP Made Delivery Nozzles
Polypropylene Nozzle and Glass Sinker - nozzle connects to the pump tubing for dispensing of liquids
Brand:  DWK Life Sciences 374326
Code : NEW
Additional Details : Weight : 0.92990kg
 View more versions of this product
Product Code. 12491531
  • £51.05 / Pack of 6
Estimated Shipment: 12-09-2024
to see stock.
Add to basket
Description
Description
  • Glass sinker is used to secure tubing when drawing from a container
Specifications
Specifications
Delivery Nozzle
Autoclavable
Wheaton peristaltic pumps, dispensing pumps and tubing
6 mm
Polypropylene