missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Red
Shop All Wheaton Science Products ProductsDescription
- Packaged in case of 100 or 1000
Specifications
Specifications
| Color | Red |
| Material | Polypropylene |
| No. per Case | 1000 Cs. |
| Neck Finish | 13-425 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Dropper Cap |
| For Use With (Equipment) | Dropping bottle |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?