missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Gray

Product Code. p-2429807 Shop All Wheaton Science Products Products
missing translation for 'orderingAttributeHoverText'
Neck Finish:
13-425
15-425
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
missing translation for 'unitSize'
Pack of 100
Pack of 1000
This item is not returnable. View return policy

Product Code. 12406161

missing translation for 'mfr': DWK Life Sciences W242535A

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
Material Polypropylene
No. per Case 1000/Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Plastic dropping bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.