missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Gray
Shop All Wheaton Science Products Products
Click to view available options
Neck Finish:
13-425
15-425
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
Unit Size:
Pack of 100
Pack of 1000
Description
- Packaged in case of 100 or 1000
Specifications
Specifications
| Color | Gray |
| Material | Polypropylene |
| No. per Case | 100/Cs. |
| Neck Finish | 8-425 |
| Certifications/Compliance | ISO 9001 |
| Product Type | Polypropylene Cap |
| For Use With (Equipment) | Plastic dropping bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction