missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for S/T Bottles

Product Code. 11784229 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Black
White
Product Type:
Cap
Open Top Cap
Unit Size:
Pack of 300
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Product Type unitSize
11784229 Black Open Top Cap Pack of 300
11794229 Black Cap Pack of 300
11581235 White Open Top Cap Pack of 300
11511105 White Cap Pack of 300
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11784229 Supplier DWK Life Sciences Supplier No. 24071601
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with Wheaton Glass Diagnostic Bottles

  • Unlined
  • Open- and closed-top caps
  • 20-400 screw thread
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Polypropylene
No. per Case 300/Cs.
Neck Finish 20-400
Certifications/Compliance ISO 9001
Product Type Open Top Cap
For Use With (Equipment) Screw neck diagnostic bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.