missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles
Shop All Wheaton Science Products Products
Click to view available options
Color:
Blue
Red
White
Includes:
Inner Sealing Ring
No Sealing Ring
PTFE/Silicone Liner
Unit Size:
Pack of 12
Description
- 45mm screw thread
- Polypropylene
- Autoclavable
- Linerless
Specifications
Specifications
| Color | Red |
| Includes | Inner Sealing Ring |
| Material | Polypropylene |
| No. per Case | 12 Cs. |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | Lab 45 graduated bottle |
| Autoclavable | Autoclavable |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction