missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences™ Wheaton™ Polyacetal Clamps

Product Code. 12458922 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Blue
Dark Green
Yellow
Unit Size:
Pack of 12
Product Code. Color unitSize
12458922 Dark Green Pack of 12
12438922 Yellow Pack of 12
12448922 Blue Pack of 12
3 options
This item is not returnable. View return policy

Product Code. 12458922

Brand: DWK Life Sciences™ 297752

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color-coded clamps hold standard taper joints securely under moderate pressure

  • Molded from polyoxymethylene (POM), an acetal resin
  • Will not scratch glass and can withstand temperatures up to 150°C
  • Resistant to bases and weak acids
  • Not resistant to halogen gases and concentrated acids
TRUSTED_SUSTAINABILITY

Specifications

Type Standard Taper Clamp
Material POM Acetal
For Use With (Equipment) Wheaton Sulfide Distillation Kit
Color Dark Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.