missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™PIPET-PAL Pipet Controller

Product Code. 17526255
Change view
Click to view available options
Volume (Metric):
1 to 100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
17526255 1 to 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17526255 Supplier DWK Life Sciences Supplier No. W155091
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Designed to be comfortable during prolonged pipetting

  • Lightweight and well-balanced
  • Quiet, robust pump for high pipetting speed
  • Precise finger-control of pipetting enables accurate dispensing, even drop-by-drop
  • Versatile for both fine dosing and gravimetric dispensing
  • Designed for use with all serological pipettes in the volume range 1-100mL
  • Eliminates contamination with a replaceable, sterile filter Lithium-Polymer battery technology for long operating time
  • Autoclavable pipette holder for cleanliness and safety
  • Supplied with a universal power supply for worldwide use
TRUSTED_SUSTAINABILITY

Specifications

Battery Type Li-polymer
Autoclavable Autoclavable
Volume (Metric) 1 to 100 mL
For Use With (Application) Serological Pipetting
Weight (English) 0.42 lb.
Weight (Metric) 195 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.