Learn More
DWK Life Sciences Wheaton™ M-T Vial File™ and Accessories
For compact and organized sample storage
Brand:  DWK Life Sciences W228786
Additional Details : Weight : 0.45000kg
 View more versions of this product
Product Code. 15843731
  • £28.60 / Pack of 6
Estimated Shipment: 18-10-2024
to see stock.
Description
Description
For compact and organized sample storage
  • Stores vials in clear polystyrene case
  • Foam inserts and partitions provide protection
  • With alphanumerical indexing card for identification
  • Index cards and partitions also available separately
Specifications
Specifications
Storage and Transportation of Sample Vials
W228790 Hinged Case
Partition
6 Case