missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Lyophilized Stopper: 2-Leg Lyophilization Stopper

Product Code. 11501105 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Gray
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11501105 Gray Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11501105 Supplier DWK Life Sciences Supplier No. W224100406
Add to basket
Add to basket
This item is not returnable. View return policy

Very low adsorption of the preservatives present in aqueous parenteral solutions - Available in 2-Leg, 3-Leg and Igloo styles.

  • Demonstrates a very low level of moisture adsorption
  • Autoclavable
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) Inner 1.3 cm
Diameter (Metric) 2 cm
For Use With (Application) Lyophilization
Hardness 46 Durometer
Autoclavable Autoclavable
Color Gray
Material Bromobutyl Rubber
Product Type 2-Leg Lyophilization Stopper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.