missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips

Product Code. 10438063 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material No. per Case unitSize
10438063 Yellow LDPE 100 Cs. Pack of 100
12436021 Blue Polyethylene 1000 Cs. Pack of 1000
10697333 Blue LDPE 100 Cs. Pack of 100
12456021 Gray Polyethylene 1000 Cs. Pack of 1000
12438782 Gray LDPE 100 Cs. Pack of 100
12476021 Green Polyethylene 1000 Cs. Pack of 1000
12448782 Green LDPE 100 Cs. Pack of 100
10285702 Natural LDPE 1000 Cs. Pack of 1000
12416021 Orange Polyethylene 1000 Cs. Pack of 1000
12418782 Orange LDPE 100 Cs. Pack of 100
12476011 Pink LDPE 1000 Cs. Pack of 1000
12488772 Pink LDPE 100 Cs. Pack of 100
12496021 Purple Polyethylene 1000 Cs. Pack of 1000
11734209 Purple LDPE 100 Cs. Pack of 100
10348773 Red Polyethylene 1000 Cs. Pack of 1000
10082643 Red LDPE 100 Cs. Pack of 100
10488623 White LDPE 100 Cs. Pack of 100
12416031 Yellow Polyethylene 1000 Cs. Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
This item is not returnable. View return policy
Product Code. 10438063 Supplier DWK Life Sciences Supplier No. 242408

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

8mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Yellow
Material LDPE
No. per Case 100 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.