missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Lab 45 Graduated Safety-Coated Bottles

Product Code. p-43398 Shop All Wheaton Science Products Products
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 12389899

Brand: DWK Life Sciences W219937

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use as media or reagent bottle

Plastisol coated for safety

  • Made from clear borosilicate glass
  • Attached 45mm white polypropylene screw cap
  • Pour ring to eliminate drips
  • Autoclavable
  • Color-coded caps available separately

TRUSTED_SUSTAINABILITY

Specifications

Capacity 8.5 oz., 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.5 oz.
Color Clear
Diameter (English) 3.97 in.
Includes Wedge shaped inner sealing ring and Polypropylene pouring ring
Material Borosilicate Glass
Closure Color White
Closure Included Yes
Graduated Yes
Height (Metric) 233 mm
Height (English) 9.17 in.
Closure Material Polypropylene
Closure Size 45 mm
Diameter (Metric) 101 mm
Closure Type Screw Cap
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.