missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Lab 45 Graduated Safety-Coated Bottles

Product Code. 12389899 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Unit Size:
Pack of 12
Product Code. Capacity (Metric) unitSize
12389899 250 mL Pack of 12
10773544 500 mL Pack of 12
12412541 1000 mL Pack of 12
3 options
This item is not returnable. View return policy

Product Code. 12389899

Brand: DWK Life Sciences W219937

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Use as media or reagent bottle

Plastisol coated for safety

  • Made from clear borosilicate glass
  • Attached 45mm white polypropylene screw cap
  • Pour ring to eliminate drips
  • Autoclavable
  • Color-coded caps available separately

TRUSTED_SUSTAINABILITY

Especificaciones

Capacity 8.5 oz., 250mL
Capacity (Metric) 250 mL
Capacity (English) 8.5 oz.
Color Clear
Diameter (English) 3.97 in.
Includes Wedge shaped inner sealing ring and Polypropylene pouring ring
Material Borosilicate Glass
Closure Color White
Closure Included Yes
Graduated Yes
Height (Metric) 233 mm
Height (English) 9.17 in.
Closure Material Polypropylene
Closure Size 45 mm
Diameter (Metric) 101 mm
Closure Type Screw Cap
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.