Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
£170.00 - £249.00
Specifications
Includes | Top cap with liners, glass rod, magnet holder, magnet and stirring paddle |
---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
10168611
|
DWK Life Sciences
356904 |
1000mL Celstir™ Spinner Flask |
£219.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
12431421
|
DWK Life Sciences
356896 |
125mL Celstir™ Spinner Flask |
£191.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
12441421
|
DWK Life Sciences
356902 |
500mL Celstir™ Spinner Flask |
£225.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11738869
|
DWK Life Sciences
356899 |
250mL Celstir™ Spinner Flask |
£191.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
12421421
|
DWK Life Sciences
356895 |
50mL Celstir™ Spinner Flask |
£170.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
10178611
|
DWK Life Sciences
356907 |
3000mL Celstir™ Spinner Flask |
£249.00
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
Description
- This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle |