Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
Product Code. 10168611 Shop All Wheaton Science Products Products
Change view
Click to view available options
For Use With (Equipment):
1000mL Celstir Spinner Flask
125mL Celstir Spinner Flask
250mL Celstir Spinner Flask
3000mL Celstir Spinner Flask
500mL Celstir Spinner Flask
50mL Celstir Spinner Flask
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10168611 1000mL Celstir Spinner Flask Each
12431421 125mL Celstir Spinner Flask Each
12441421 500mL Celstir Spinner Flask Each
11738869 250mL Celstir Spinner Flask Each
12421421 50mL Celstir Spinner Flask Each
10178611 3000mL Celstir Spinner Flask Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10168611 Supplier DWK Life Sciences Supplier No. 356904
Only null left
Add to basket
Add to basket
Replacement for Double Sidearm Celstir and Jacketed Double Sidearm Celstir.
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) 1000mL Celstir Spinner Flask
Includes Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
Type Impeller