missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Eliminate use of separate aluminum seal and rubber stopper
- Polypropylene closures
- Gray bromobutyl stoppers
Specifications
Specifications
| Material (Liner) | Bromobutyl |
| Color | White, Gray |
| Material | Polypropylene |
| Closure Size | 20 mm |
| For Use With (Equipment) | 20-400 Screw Thread Vials |
| Autoclavable | Autoclavable |
| Product Type | Screw Cap |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?