missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ I-LOC Closures

Product Code. 11788709 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Black
Blue
Red
White, Gray
Yellow
Unit Size:
Pack of 100
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11788709 Black Pack of 100
12498722 Red Pack of 100
11551235 White, Gray Pack of 100
12418732 Yellow Pack of 100
12408732 Blue Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11788709 Supplier DWK Life Sciences Supplier No. 24067601
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Eliminate use of separate aluminum seal and rubber stopper

  • Polypropylene closures
  • Gray bromobutyl stoppers

TRUSTED_SUSTAINABILITY

Specifications

Material (Liner) Bromobutyl
Color Black
Material Polypropylene
Closure Size 20 mm
For Use With (Equipment) 20-400 Screw Thread Vials
Autoclavable Autoclavable
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.