Learn More
DWK Life Sciences Wheaton™ Hybridization Bottles
For use in nucleic acid and protein blotting procedures that require a minimum of probe solution
Brand: DWK Life Sciences 805001
73.25 GBP valid until 2024-12-31
Use promo code "23261" to get your promotional price.
Additional Details : Weight : 1.38350kg
Description
- Manufactured from heavy-wall borosilicate glass
- Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
- Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
- Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage
Order Info
Items come as one per case
Specifications
Polycarbonate | |
Hybridization Bottle | |
Nucleic acid and protein blotting procedures that require a minimum of probe solution | |
Screw | |
35 mm | |
45 mm | |
100 mm |
3.93 in. | |
1.37 in. | |
Borosilicate Glass | |
PTFE or Silicone | |
Plastisol | |
USP Type I |