missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ HDPE Serum Bottles
Click to view available options
Capacity (Metric):
15 mL
30 mL
60 mL
120 mL
Diameter (English):
1.1 in.
1.25 in.
1.49 in.
1.85 in.
Height (English):
2.2 in.
2.63 in.
3.3 in.
4.05 in.
Description
- Manufactured from high density polyethylene
- Widely used for animal health products and storage of non-medical products
- Accepts standard rubber stoppers and aluminum seals
Specifications
Capacity | 0.5 oz., 15 mL |
Closure Material | Rubber |
Diameter (Metric) | 28 mm |
Height (English) | 2.2 in. |
Height (Metric) | 56 mm |
Product Type | Serum Bottle |
Capacity (Metric) | 15 mL |
Capacity (English) | 0.5 oz. |
Color | Natural |
Diameter (English) | 1.1 in. |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'