missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Extended Controlled 20mm LDPE Dropper Tips

Product Code. 12436081 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Blue
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
Unit Size:
Pack of 100
Pack of 1000
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Material No. per Case unitSize
12436081 Blue Polyethylene 1000 Cs. Pack of 1000
10793504 Blue LDPE 100 Cs. Pack of 100
12476081 Green Polyethylene 1000 Cs. Pack of 1000
12448812 Green LDPE 100 Cs. Pack of 100
12456071 Natural Polyethylene 1000 Cs. Pack of 1000
12416081 Orange Polyethylene 1000 Cs. Pack of 1000
12418812 Orange LDPE 100 Cs. Pack of 100
12476071 Pink Polyethylene 1000 Cs. Pack of 1000
12496081 Purple Polyethylene 1000 Cs. Pack of 1000
12496071 Red Polyethylene 1000 Cs. Pack of 1000
12408812 Red LDPE 100 Cs. Pack of 100
12436091 White Polyethylene 1000 Cs. Pack of 1000
11704219 White LDPE 100 Cs. Pack of 100
12416091 Yellow Polyethylene 1000 Cs. Pack of 1000
12468812 Yellow LDPE 100 Cs. Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Product Code. 12436081 Supplier DWK Life Sciences Supplier No. W242434A
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

20mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Material Polyethylene
No. per Case 1000 Cs.
Certifications/Compliance ISO 9001
Drop Size 41 μL
Product Type Dropper Tip
For Use With (Equipment) Plastic dropping bottle
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.