Learn More
DWK Life Sciences Wheaton™ E-Z Seals With PTFE/Grey Butyl Liner
Product Code. 12472701 Shop All Wheaton Science Products Products
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
Product Code. 12472701
Brand: DWK Life Sciences 22423501
Only null left
Add to basket
Add to basket
Specifications
Material (Liner) PTFE/Butyl
Color Natural
Material Aluminum
Product Line E-Z Seal
Quantity 1000 Cs.
Type Open Top Cap
Closure Size 11 mm