Learn More
DWK Life Sciences Wheaton™ Connection Connectors
Product Code. 12425442 Shop All Wheaton Science Products Products
Click to view available options
Thread Size:
12-425 and 24-400
20-400 and 24-400
Unit Size:
Pack of 6
Product Code. 12425442
Brand: DWK Life Sciences 125482
Only null left
Add to basket
Add to basket
Safely joins two exterior threaded glass components without hooks, springs or clamps
  • Screw cap configuration creates a grease-free, vacuum- tight seal
  • Inert contact surface
  • Easy to assemble
  • Connection can be used under moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
  • Available with white polypropylene or black phenolic caps
  • Phenolic connectors have a working temperature of up to 180
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USP Type I, ASTM E438 Type I, Class A
For Use With (Application) Moderate pressure to generate a gas or deliver a reagent under an inert atmosphere
For Use With (Equipment) Micro soxhlet extraction apparatus
Includes Screw cap
Material Borosilicate Glass, Polypropylene
Product Type Screw Thread Connector
Thread Size 12-425 and 24-400