missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits

Product Code. 16237310 Shop All Wheaton Science Products Products
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
Omni Flex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Sterile
OmniFlex 3G Sterile Igloo Lyo Stopper
OmniFlex 3G Sterile Serum Stopper
Ultra Pure Sterile
Ultra Pure Sterile Serum Stopper
Color (Seal):
Blue
Red
Unit Size:
Pack of 158
Pack of 200
Pack of 210
This item is not returnable. View return policy

Product Code. 16237310

Brand: DWK Life Sciences W990012CP

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

Wheaton CompletePAK product line is a range of crimp top vials, rubber stoppers and seals that are ready-to-use and are off-the-shelf, customizable for any application

  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

TRUSTED_SUSTAINABILITY

Specifications

Color (Seal) Blue
Color Clear
Sterility Sterile
Volume (Metric) 10 mL
Includes Vial, Stopper, and Seal
No. per Case 158
Product Line Wheaton CompletePAK
Closure Type Ultra Pure Sterile Serum Stopper
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Closure Size 20 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.