missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Straight-Sided Jars with TFE-lined Caps

Product Code. 10275302 Shop All Wheaton Science Products Products
Change view
Click to view available options
Capacity (Metric):
60 mL
125 mL
250 mL
500 mL
1000 mL
Closure Size:
53-400 mm
58-400 mm
70-400 mm
89-400 mm
Unit Size:
Pack of 12
Pack of 24
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Size unitSize
10275302 1000 mL 89-400 mm Pack of 12
10632373 60 mL 53-400 mm Pack of 24
10317092 125 mL 58-400 mm Pack of 24
11591115 250 mL 70-400 mm Pack of 12
11501125 500 mL 89-400 mm Pack of 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 10275302 Supplier DWK Life Sciences Supplier No. W216912

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for soil sampling and environmental applications

Excellent for storage of dry materials and specimens, visible through clear Type III soda-lime glass

  • Walls allow for complete removal of contents
  • Wide-mouth design is great for large solid samples
  • Each case of Convenience Pack Bottles includes Hazard Communication labels for proper content identification and Tamper Evident labels to protect sample integrity in transit or long-term storage

Order Info

Available with caps attached or without caps. Caps can be ordered separately.

Compliance

Clear USP Type III soda-lime glass

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 1000 mL
Capacity (English) 32 oz.
Color Clear
Diameter (English) 3.74 in.
Material Soda Lime Glass
Closure Color White
Height (Metric) 170 mm
Height (English) 6.69 in.
Closure Liner PTFE
Closure Material Polypropylene
Closure Size 89-400 mm
Diameter (Metric) 95 mm
Closure Type Screw Cap
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.