missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with PE Cone-Lined Caps
Shop All Wheaton Science Products ProductsDescription
- Well suited for storage and sampling of liquids; clear Type III soda-lime glass provides visibility of contents
- Narrow mouth enables liquids to be carefully poured to prevent spillage
Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage.
Order Info
Caps can be ordered separately.
Specifications
Specifications
| Capacity | 1 oz., 30mL |
| Capacity (Metric) | 30 mL |
| Capacity (English) | 1 oz. |
| Color | Clear |
| Diameter (English) | 1.22 in. |
| Material | Soda Lime Glass |
| Closure Color | Black |
| Certifications/Compliance | EPA, FDA |
| Height (Metric) | 79 mm |
| Height (English) | 3.1 in. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?