missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials
Shop All Wheaton Science Products ProductsDescription
- Screw caps for WHEATON liquid scintillation vials
- Case of 1000 provides an economical alterative
- Not autoclavable
Specifications
Specifications
| Description | Cap Size: 24-400 |
| Material (Liner) | Metal Foil |
| Color | White |
| Material | Urea |
| Closure Size | 24-400 |
| For Use With (Equipment) | Liquid Scintillation Vial |
| Product Type | Screw Cap |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?