missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Caps With PE Poly-Seal Liner

Product Code. 12433881 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
13-425
18-400
28-400
38-430
No. per Case:
100 Cs.
144 Cs.
Unit Size:
Pack of 100
Pack of 144
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish No. per Case unitSize
12433881 38-430 100 Cs. Pack of 100
12413881 28-400 100 Cs. Pack of 100
11759685 18-400 144 Cs. Pack of 144
11724219 13-425 144 Cs. Pack of 144
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12433881 Supplier DWK Life Sciences Supplier No. 240181
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Unique cone liner design offers superior torque retention and excellent sealing characteristics

  • Poly-Seal liner is manufactured from low density polyethylene
  • Screw Cap
  • Phenolic
  • Color: Black

It is recommended that liner be tested prior to use for leak resistant applications

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Neck Finish 38-430
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Polyethylene
Closure Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.