missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials
Shop All Wheaton Science Products ProductsDescription
Designed to ensure the ultimate in product safety
- Autoclavable
- Liner made of styrene-butadiene rubber faced with PTFE
- Excellent chemical resistance
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 250 Cs. |
| Neck Finish | 13-425 |
| Certifications/Compliance | ISO 9001 |
| For Use With (Equipment) | 2 and 4mL Vial |
| Closure Liner | PTFE/Rubber |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?