missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Solid Top Screw Caps

Product Code. 12435951 Shop All Wheaton Science Products Products
Change view
Click to view available options
Closure Size:
15-425
18-400
20-400
24-400
Unit Size:
Pack of 144
Pack of 200
Pack of 500
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size unitSize
12435951 24-400 Pack of 144
12415081 20-400 Pack of 144
10423332 18-400 Pack of 500
10130721 15-425 Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 12435951 Supplier DWK Life Sciences Supplier No. W239300
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.

Providing safe, secure and chemically inert storage. Wheaton™ Black Phenolic Solid Top Screw Caps are manufactured from the highest quality materials.

  • 14B rubber lined caps feature a nontoxic white styrene-butadiene rubber liner ideal for cell culture work
  • Aluminum foil faced pulp board liners can be used with organic solvents
  • Use LDPE lined caps with distilled water, analytical standards and reagents
TRUSTED_SUSTAINABILITY

Specifications

Color Black
For Use With (Application) Ideal for cell culture work
Material Phenolic
Liner Material Styrene Butadiene Rubber
Product Type Screw Cap
Closure Size 24-400
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.