missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with PTFE-Faced Rubber Liner

Product Code. 10246862 Shop All Wheaton Science Products Products
Change view
Click to view available options
Neck Finish:
13-425
15-415
15-425
18-400
18-415
20-400
28-400
33-400
33-430
No. per Case:
100 Cs.
200 Cs.
Unit Size:
Pack of 100
Pack of 200
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Neck Finish No. per Case unitSize
10246862 18-400 200 Cs. Pack of 200
10630375 33-430 100 Cs. Pack of 100
10058860 15-425 200 Cs. Pack of 200
10143721 20-400 100 Cs. Pack of 100
11744219 28-400 100 Cs. Pack of 100
10555160 15-415 200 Cs. Pack of 200
11774219 33-400 100 Cs. Pack of 100
12458712 18-415 200 Cs. Pack of 200
10778571 13-425 200 Cs. Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
This item is not returnable. View return policy
Product Code. 10246862 Supplier DWK Life Sciences Supplier No. 240415
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Features PTFE faced (14B) styrene-butadiene rubber liner

  • Designed for product safety
  • TFE/Styrene-Butadiene rubber liner
  • Screw cap
  • Pheolic
  • Black
  • Autoclavable

TRUSTED_SUSTAINABILITY

Specifications

Color Black
Material Phenolic Resin
No. per Case 200 Cs.
Neck Finish 18-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner PTFE/14B Rubber
Autoclavable Autoclavable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.