missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Open-Top Screw Caps Without Liners

Product Code. 10399143 Shop All Wheaton Science Products Products
Change view
Click to view available options
Thread Size (Metric):
24 mm
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Thread Size (Metric) Unit Size
10399143 24 mm Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10399143 Supplier DWK Life Sciences Supplier No. W240506
  • ONLINE EXCLUSIVE PRICE
    £39.60
    £38.87 / Pack of 200
    Save £0.73
    1% Off
 Restricted Item
Estimated Shipment: 07-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Black
Material Phenolic Resin
No. per Case 200 Cs.
Thread Size (Metric) 24 mm
Neck Finish 8-425
For Use With (Equipment) Autosampler vial
Certifications/Compliance ISO 9001
Product Type Open Top Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.