missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Open Top Cap with Gray Butyl Septa and Flange
Click to view available options
No. per Case:
100 Case
1000 Case
Description
Replacement Black Phenolic Screw Caps for Media Bottles
- Open Top Cap With Gray Chlorobutyl / 50 Septa and Flange
- Autoclavable
Specifications
Specifications
Color | Black |
Includes | Gray Butyl septa and flange |
Material | Phenolic Resin |
No. per Case | 100 Case |
Neck Finish | 33-430 |
Certifications/Compliance | ISO 9001 |
Product Type | Open Top Cap |
For Use With (Equipment) | Bottle |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'