missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Black Phenolic Linerless Screw Caps

Product Code. 10021432 Shop All Wheaton Science Products Products
Change view
Click to view available options
Color:
Black
Unit Size:
Pack of 200
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
10021432 Black Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10021432 Supplier DWK Life Sciences Supplier No. W240518
  • ONLINE EXCLUSIVE PRICE
    £125.76
    £109.56 / Pack of 200
    Save £16.20
    12% Off
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

WHEATON™ Black Phenolic Linerless Screw Caps feature an open top design that allows easy syringe access. These replacement 24-400 phenolic caps are supplied without septa and are sold in cases of 200, making them a practical choice for high-volume laboratory workflows.

  • Black phenolic open top screw cap without septa
  • Open top design provides direct syringe access
  • Compatible with 24-400 threaded vials and containers
  • Supplied as replacement caps for EPA and laboratory vials
  • Available in cases of 200 for high-volume use

Specifications

Color Black
Material Phenolic Resin
Sterility Non-sterile
For Use With (Equipment) Bottle
Closure Size 24-400
Autoclavable Autoclavable
Disposable Reusable
Product Type Screw Cap
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.