Learn More
DWK Life Sciences Wheaton™ Center Disc Tear-Out Aluminum Seals, Unlined
Product Code. 12482671 Shop All Wheaton Science Products Products
Click to view available options
Color:
Natural
Unit Size:
Pack of 1000
Product Code. 12482671
Brand: DWK Life Sciences 224189
Only null left
Add to basket
Add to basket
Tear-out seals remove completely allowing content to pour from bottle. Wheaton™ Center Disc Tear-Our Aluminum Seals, Unlined are available in a wide range of sizes and colors.
  • Good barrier properties
  • Color selection includes natural, red, blue and green
  • Chemically resistant to alcohol and acetone, among others
TRUSTED_SUSTAINABILITY
Specifications
Color Natural
Material Aluminum
Quantity 1000/Cs.
Type Solid Top Cap
Closure Size 11 mm