missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ TurboBlotter Transfer System, Small Kits
Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA
£117.97 - £236.00
Specifications
For Use With (Application) | DNA/RNA Transfer |
---|---|
Type | Nytran™ SPC 0.45μm TurboBlotter™ Kit |
Includes | 12 x 16cm Small transfer device, 5 sets of transfer materials (set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o |
Includes: Includes device plus five sets of transfer materials for gel size indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Price | Quantity & Availability | ||||
10325561
|
Cytiva
10416304 |
11 x 14 cm Gel TurboBlotter™ Transfer System | Nytran™ SPC 0.45μm TurboBlotter™ Kit |
£230.00
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
||||
11989287
|
Cytiva
10416300 |
10 x 15 cm Gel TurboBlotter™ Transfer System | Nytran™ SPC 0.45μm TurboBlotter™ Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
11929307
|
Cytiva
10416328 |
7 x 10 cm Gel TurboBlotter™ Transfer System | Nytran™ SPC 0.45μm TurboBlotter™ Kit |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
- Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
- Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
- Very easy to set up and works without power or vacuum source
- Requires less buffer and blotting paper than conventional transfers
- Reusable
- Model is 12 × 16cm, for gels from 7 × 8cm to 11 × 14cm
Order Info
Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).
Specifications
DNA/RNA Transfer | |
Nytran™ SPC 0.45μm TurboBlotter™ Kit | |
12 x 16cm Small transfer device, 5 sets of transfer materials (set consists of one sheet of Nytran™ SPC 0.45μm Nylon membrane, one wick of 3mm chromatography blotting Paper and a blotter stack composed of eight sheets o |