missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ TurboBlotter Transfer System, Large Kits

Product Code. 11969297
Click to view available options
Length (English) Gel:
7.8 in.
9.8 in.
Length (Metric) Gel:
20 cm
25 cm
Width (English) Gel:
5.9 in.
7.8 in.
Width (Metric) Gel:
15 cm
20 cm
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11969297

Brand: Cytiva 10416316

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Includes

Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytrantrademark (TM) SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.

Downward blotting device turns the conventional Northern/Southern transfer stack upside down for rapid, high-resolution transfer of DNA and RNA

  • Takes advantage of gravity — no heavy weights are required on the top of the transfer stack, eliminating the messy set-up of standard upward capillary transfers
  • Has a smaller footprint than most homemade devices and is stackable — up to five units can be stacked during transfers
  • Allows for alkaline buffer transfers in one hour and neutral (SSC) transfers in three hours
  • Very easy to set up and works without power or vacuum source
  • Requires less buffer and blotting paper than conventional transfers
  • Reusable

  • Model is 21 × 26cm, for gels from 12 × 21cm to 20 × 25cm

Order Info

Refill packs containing additional membranes and papers required for further transfer are available separately (Cat. Nos. 09-301-177, -179, -181, -183, -185 and -189).

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) 15 x 20 cm Gel TurboBlotter™ Transfer System
Type Nytran™ SPC 0.45μm TurboBlotter™ Kit
Includes Includes device plus five sets of transfer materials for gel size (21 x 26cm) indicated in ordering table. Each set consists of one sheet of Nytran™ SPC 0.45Glc mum Nylon membrane, one wick of 3MM Chr blotting paper and a blotter stack composed of eight sheets of 3MM Chr blotting paper and 20 sheets of GB004 blotting paper. Refills, available separately, contain five sets of transfer materials.
Length (English) Gel 7.8 in.
Length (Metric) Gel 20 cm
Width (English) Gel 5.9 in.
Width (Metric) Gel 15 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato