Learn More
Cytiva Whatman™ Syringe-Type Membrane Holder
Stainless steel membrane holder for use with a standard syringe
Brand:  Cytiva 1980-001
Code : 75
Additional Details : Weight : 0.06000kg
 View more versions of this product
Product Code. 11370644
  • £167.00 / Each
Estimated Shipment: 16-05-2024
to see stock.
Add to basket
Description
Description
  • Designed for the quick and easy clarification, sterilization, and removal of particulates from small volume samples (typically for HPLC applications)
  • holders contain PTFE gaskets and O-rings and allow the membrane to be autoclaved in place without the filter sticking to the holder.
  • Luer lock fittings connect to a standard syringe and offer convenience and ease of use for clarification, sterilization, and removal of particulates from small volumes of liquid
Specifications
Specifications
Stainless Steel
13 mm