Learn More
Qiagen SPOT CloneSaver Holder
Product Code. 11393415
Click to view available options
:
Rigid Frame
Product Code. 11393415
Brand: Qiagen WB100034
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
SPOT CloneSaver holder
SPOT CloneSaver Holder is a rigid frame that allows automated spotting to standard CloneSaver Cards. It keeps the card flat for uniform and precise spotting of biological samples. The 96-well card is easily inserted into the ANSI/SBS-compatible frame, which can then be placed onto a liquid-handling deck just like a multiwell plate.
The SPOT CloneSaver Holder is compatible with standard liquid handlers manufactured by companies such as Beckman Coulter and Tecan Instruments.
Specifications
Product Type CloneSaver Card Holder
Description Keeps card flat for uniform, precise spotting of biological samples
For Use With (Equipment) Compatible w/standard liquid handlers